Search
Search

MAP70-5| Gene Symbol: MAP70-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117827.3 | NP_193455.2 | 5 - 6 | 874 | 65 | |
| GenBank mRNA | AK117336.1 | - | 5 - 6 | 788 | 65 | |
| AM086442.1 | - | 5 - 6 | 761 | 65 | ||
| BT005901.1 | - | 5 - 6 | 703 | 65 | ||
Set Membership: |
|