Search
Search

SS4| Gene Symbol: SS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117934.4 | NP_193558.3 | 15 - 16 | 3060 | 132 | |
| GenBank mRNA | AK226610.1 | - | 15 - 16 | 3065 | 132 | |
| KJ676990.1 | - | 15 - 16 | 2947 | 132 | ||
Set Membership: |
|