Search
Search

BIN2| Gene Symbol: BIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117987.4 | NP_193606.1 | 8 - 9 | 1193 | 92 | |
| GenBank mRNA | AY086529.1 | - | 8 - 9 | 1029 | 92 | |
| BT026031.1 | - | 7 - 8 | 857 | 92 | ||
Set Membership: |
|