Search
Search

HEXO3| Gene Symbol: HEXO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105233.5 | NP_176737.2 | 3 - 4 | 898 | 134 | |
| GenBank mRNA | AY128283.1 | - | 3 - 4 | 472 | 134 | |
| BT000831.1 | - | 3 - 4 | 454 | 134 | ||
Set Membership: |
|