Search
Search

HEXO3| Gene Symbol: HEXO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105233.5 | NP_176737.2 | 6 - 7 | 1212 | 86 | |
| GenBank mRNA | AY128283.1 | - | 6 - 7 | 786 | 86 | |
| BT000831.1 | - | 6 - 7 | 768 | 86 | ||
Set Membership: |
|