Search
Search

AT1G65920| Gene Symbol: AT1G65920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334226.1 | NP_001320309.1 | 2 - 3 | 681 | 101 | |
| NM_105264.3 | NP_176767.1 | 3 - 4 | 387 | 101 | ||
Set Membership: |
|