Search
Search

REV3| Gene Symbol: REV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334301.1 | NP_001319333.1 | 3 - 4 | 322 | 76 | |
| NM_001334302.1 | NP_001320390.1 | 3 - 4 | 373 | 76 | ||
| NM_001334303.1 | NP_001320389.1 | 3 - 4 | 354 | 76 | ||
| NM_001334304.1 | NP_001320393.1 | 3 - 4 | 350 | 76 | ||
| NM_001334305.1 | NP_001320391.1 | 3 - 4 | 349 | 76 | ||
| NM_001334306.1 | NP_001320392.1 | 3 - 4 | 363 | 76 | ||
| NM_105417.4 | NP_176917.2 | 3 - 4 | 322 | 76 | ||
| GenBank mRNA | AB114052.1 | - | 2 - 3 | 190 | 76 | |
Set Membership: |
|