Search
Search

PHO1;H1| Gene Symbol: PHO1;H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334380.1 | NP_001322237.1 | 5 - 6 | 1576 | 73 | |
| NM_105547.3 | NP_564940.1 | 5 - 6 | 1576 | 73 | ||
| GenBank mRNA | AY057574.1 | - | 5 - 6 | 1250 | 73 | |
Set Membership: |
|