Search
Search

BCA4| Gene Symbol: BCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105709.4 | NP_177198.1 | 7 - 8 | 838 | 62 | |
| NM_179541.3 | NP_849872.1 | 5 - 6 | 1337 | 62 | ||
| NM_202390.3 | NP_974119.1 | 6 - 7 | 1247 | 62 | ||
| GenBank mRNA | AY034926.1 | - | 7 - 8 | 620 | 62 | |
| AY087827.1 | - | 7 - 8 | 636 | 62 | ||
| AY113922.1 | - | 6 - 7 | 482 | 62 | ||
Set Membership: |
|