Search
Search

CSN5B| Gene Symbol: CSN5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334491.1 | NP_001321294.1 | 6 - 7 | 1010 | 85 | |
| NM_105792.2 | NP_177279.1 | 5 - 6 | 1143 | 85 | ||
| GenBank mRNA | AF395061.1 | - | 5 - 6 | 989 | 85 | |
| AF411778.1 | - | 5 - 6 | 1088 | 85 | ||
| AY124816.1 | - | 5 - 6 | 989 | 85 | ||
Set Membership: |
|