Search
Search

ATWHY2| Gene Symbol: ATWHY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105795.5 | NP_177282.2 | 1 - 2 | 98 | 77 | |
| GenBank mRNA | AB493532.1 | - | 1 - 2 | 35 | 77 | |
| AY072110.1 | - | 1 - 2 | 62 | 77 | ||
| AY122961.1 | - | 1 - 2 | 35 | 77 | ||
Set Membership: |
|