Search
Search

CK1| Gene Symbol: CK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105828.4 | NP_177315.1 | 6 - 7 | 1511 | 157 | |
| GenBank mRNA | AK229787.1 | - | 6 - 7 | 1149 | 157 | |
| AK230407.1 | - | 6 - 7 | 1148 | 157 | ||
| BT008311.1 | - | 6 - 7 | 942 | 157 | ||
Set Membership: |
|