Search
Search

SEC6| Gene Symbol: SEC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105840.4 | NP_565026.1 | 9 - 10 | 977 | 69 | |
| GenBank mRNA | AY046014.1 | - | 9 - 10 | 898 | 69 | |
| AY133819.1 | - | 8 - 9 | 629 | 69 | ||
Set Membership: |
|