Search
Search

BGAL17| Gene Symbol: BGAL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036196.2 | NP_001031273.1 | 1 - 2 | 488 | 115 | |
| NM_001334560.1 | NP_001321343.1 | 2 - 3 | 353 | 115 | ||
| NM_105957.3 | NP_565051.1 | 1 - 2 | 488 | 115 | ||
| GenBank mRNA | AY058198.1 | - | 1 - 2 | 390 | 115 | |
| AY142008.1 | - | 1 - 2 | 169 | 115 | ||
Set Membership: |
|