Search
Search

BGLU42| Gene Symbol: BGLU42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036898.2 | NP_001031975.1 | 9 - 10 | 1171 | 113 | |
| NM_123047.4 | NP_198505.2 | 9 - 10 | 1172 | 113 | ||
| GenBank mRNA | AK175760.1 | - | 9 - 10 | 1172 | 113 | |
| BT010611.1 | - | 9 - 10 | 1032 | 113 | ||
Set Membership: |
|