Search
Search

ARF8| Gene Symbol: ARF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085203.1 | NP_001078672.1 | 9 - 10 | 1046 | 88 | |
| NM_001344162.1 | NP_001331077.1 | 8 - 9 | 1574 | 88 | ||
| NM_123060.3 | NP_198518.1 | 9 - 10 | 1621 | 88 | ||
| GenBank mRNA | AF042196.1 | - | 9 - 10 | 1046 | 88 | |
| AY669790.1 | - | 9 - 10 | 923 | 88 | ||
Set Membership: |
|