Search
Search

CIP4| Gene Symbol: CIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344168.1 | NP_001331331.1 | 2 - 3 | 310 | 112 | |
| NM_001344169.1 | NP_001331332.1 | 2 - 3 | 267 | 112 | ||
| NM_123078.3 | NP_568546.1 | 2 - 3 | 310 | 112 | ||
Set Membership: |
|