Search
Search

WSD1| Gene Symbol: WSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344178.1 | NP_001331811.1 | 3 - 4 | 927 | 103 | |
| NM_001344179.1 | NP_001331810.1 | 3 - 4 | 927 | 103 | ||
| NM_001344180.1 | NP_001331809.1 | 3 - 4 | 927 | 103 | ||
| NM_123089.3 | NP_568547.1 | 3 - 4 | 857 | 103 | ||
| GenBank mRNA | AY056316.1 | - | 3 - 4 | 848 | 103 | |
| BT015776.1 | - | 3 - 4 | 839 | 103 | ||
Set Membership: |
|