Search
Search

AT3G43610| Gene Symbol: AT3G43610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339100.1 | NP_001325537.1 | 4 - 5 | 1534 | 113 | |
| NM_001339101.1 | NP_001325538.1 | 4 - 5 | 1534 | 113 | ||
| NM_114229.3 | NP_189947.2 | 4 - 5 | 1534 | 113 | ||
Set Membership: |
|