Search
Search

GH9A4| Gene Symbol: GH9A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339113.1 | NP_001327805.1 | 2 - 3 | 667 | 64 | |
| NM_114254.3 | NP_189972.2 | 2 - 3 | 667 | 64 | ||
| GenBank mRNA | AY072099.1 | - | 2 - 3 | 627 | 64 | |
Set Membership: |
|