Search
Search

DCL3| Gene Symbol: DCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161190.2 | NP_001154662.2 | 13 - 14 | 1899 | 87 | |
| NM_001161191.3 | NP_001154663.2 | 13 - 14 | 2017 | 87 | ||
| NM_001339114.1 | NP_001325812.1 | 13 - 14 | 2027 | 87 | ||
| NM_001339115.1 | NP_001325811.1 | 13 - 14 | 2030 | 87 | ||
| NM_001339116.1 | NP_001325813.1 | 8 - 9 | 1400 | 87 | ||
| NM_114260.1 | NP_189978.1 | 11 - 12 | 1752 | 87 | ||
| GenBank mRNA | DQ479972.1 | - | 11 - 12 | 1752 | 87 | |
Set Membership: |
|