Search
Search

CR88| Gene Symbol: CR88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126439.4 | NP_178487.1 | 14 - 15 | 1904 | 88 | |
| NM_179601.1 | NP_849932.1 | 14 - 15 | 1749 | 88 | ||
| GenBank mRNA | AF436826.1 | - | 14 - 15 | 1758 | 88 | |
| AX765864.1 | - | 14 - 15 | 1675 | 88 | ||
| AY053403.1 | - | 14 - 15 | 1743 | 88 | ||
| BT002234.1 | - | 14 - 15 | 1675 | 88 | ||
Set Membership: |
|