Search
Search

NUDT5| Gene Symbol: NUDT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335236.1 | NP_001324657.1 | 8 - 9 | 1054 | 108 | |
| NM_126476.3 | NP_178524.2 | 7 - 8 | 732 | 108 | ||
| GenBank mRNA | BT010760.1 | - | 7 - 8 | 732 | 108 | |
| BT011326.1 | - | 7 - 8 | 689 | 108 | ||
Set Membership: |
|