Search
Search

AT2G05540| Gene Symbol: AT2G05540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126577.5 | NP_178622.1 | 1 - 2 | 217 | 86 | |
| GenBank mRNA | AY050337.1 | - | 1 - 2 | 187 | 86 | |
| AY052202.1 | - | 1 - 2 | 180 | 86 | ||
| AY094047.1 | - | 1 - 2 | 106 | 86 | ||
Set Membership: |
|