Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GRF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335301.1 | NP_001325112.1 | 2 - 3 | 582 | 104 | |
NM_001335302.1 | NP_001325111.1 | 1 - 2 | 612 | 104 | ||
NM_126630.2 | NP_027759.1 | 2 - 3 | 519 | 104 | ||
GenBank mRNA | AY060586.1 | - | 2 - 3 | 487 | 104 | |
AY125572.1 | - | 2 - 3 | 418 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |