Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335345.1 | NP_001323939.1 | 16 - 17 | 1667 | 116 | |
NM_001335346.1 | NP_001323940.1 | 16 - 17 | 1667 | 116 | ||
NM_001335347.1 | NP_001323936.1 | 15 - 16 | 1584 | 116 | ||
NM_001335348.1 | NP_001323937.1 | 15 - 16 | 1513 | 116 | ||
NM_001335349.1 | NP_001323938.1 | 13 - 14 | 1444 | 116 | ||
NM_126770.8 | NP_178811.7 | 15 - 16 | 1524 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |