Search
Search

ALD1| Gene Symbol: ALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126957.2 | NP_565359.1 | 9 - 10 | 1102 | 100 | |
| GenBank mRNA | AY057526.1 | - | 9 - 10 | 1073 | 100 | |
| AY143898.1 | - | 9 - 10 | 1030 | 100 | ||
| AY518702.1 | - | 9 - 10 | 1030 | 100 | ||
Set Membership: |
|