Search
Search

ALDH6B2| Gene Symbol: ALDH6B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084418.1 | NP_001077887.1 | 3 - 4 | 364 | 77 | |
| NM_001084419.1 | NP_001077888.1 | 4 - 5 | 564 | 77 | ||
| NM_126989.2 | NP_179032.1 | 4 - 5 | 564 | 77 | ||
Set Membership: |
|