Search
Search

PHB2| Gene Symbol: PHB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100266.3 | NP_171882.1 | 3 - 4 | 522 | 62 | |
| NM_202026.2 | NP_973755.1 | 2 - 3 | 480 | 62 | ||
| NM_202027.2 | NP_973756.1 | 3 - 4 | 525 | 62 | ||
| GenBank mRNA | AF370317.1 | - | 3 - 4 | 391 | 62 | |
| AY063102.1 | - | 2 - 3 | 289 | 62 | ||
| U66592.1 | - | 3 - 4 | 383 | 62 | ||
| U89791.1 | - | 3 - 4 | 372 | 62 | ||
Set Membership: |
|