Search
Search

CRT3| Gene Symbol: CRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100718.5 | NP_563816.1 | 7 - 8 | 1022 | 69 | |
| NM_202064.1 | NP_973793.1 | 7 - 8 | 728 | 69 | ||
| GenBank mRNA | AY056320.1 | - | 7 - 8 | 890 | 69 | |
Set Membership: |
|