Search
Search

AAPT1| Gene Symbol: AAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332070.1 | NP_001323307.1 | 8 - 9 | 530 | 108 | |
| NM_101226.5 | NP_172813.1 | 10 - 11 | 899 | 108 | ||
| NM_202088.2 | NP_973817.1 | 9 - 10 | 796 | 108 | ||
| GenBank mRNA | AF091843.1 | - | 10 - 11 | 736 | 108 | |
| AY080725.1 | - | 10 - 11 | 773 | 108 | ||
| AY114062.1 | - | 9 - 10 | 637 | 108 | ||
Set Membership: |
|