Search
Search

WNK1| Gene Symbol: WNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035560.1 | NP_001030637.1 | 5 - 6 | 1163 | 69 | |
| NM_001125104.1 | NP_001118576.1 | 4 - 5 | 1136 | 69 | ||
| NM_111363.4 | NP_187142.1 | 5 - 6 | 1264 | 69 | ||
| GenBank mRNA | AB084266.1 | - | 5 - 6 | 716 | 69 | |
Set Membership: |
|