Search
Search

NHX2| Gene Symbol: NHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337553.1 | NP_001319475.1 | 7 - 8 | 1059 | 120 | |
| NM_001337554.1 | NP_001326211.1 | 7 - 8 | 1306 | 120 | ||
| NM_001337555.1 | NP_001326210.1 | 6 - 7 | 1477 | 120 | ||
| NM_111375.4 | NP_187154.1 | 7 - 8 | 1306 | 120 | ||
| GenBank mRNA | AF490586.1 | - | 6 - 7 | 690 | 120 | |
Set Membership: |
|