Search
Search

ALY2| Gene Symbol: ALY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035566.2 | NP_001030643.1 | 16 - 17 | 2839 | 89 | |
| NM_001125108.2 | NP_001118580.1 | 15 - 16 | 2925 | 89 | ||
| NM_001202883.1 | NP_001189812.1 | 16 - 17 | 2804 | 89 | ||
| NM_001202884.1 | NP_001189813.1 | 17 - 18 | 2772 | 89 | ||
| NM_111411.5 | NP_187189.2 | 16 - 17 | 2836 | 89 | ||
| GenBank mRNA | AJ583496.1 | - | 16 - 17 | 2776 | 89 | |
Set Membership: |
|