Search
Search

ACBP4| Gene Symbol: ACBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111415.5 | NP_187193.3 | 13 - 14 | 1444 | 85 | |
| NM_202498.2 | NP_974227.1 | 13 - 14 | 1554 | 85 | ||
| GenBank mRNA | AK228046.1 | - | 13 - 14 | 1445 | 85 | |
| BT006458.1 | - | 13 - 14 | 1342 | 85 | ||
Set Membership: |
|