Search
Search

PLDP2| Gene Symbol: PLDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111436.6 | NP_187214.2 | 7 - 8 | 1404 | 74 | |
| GenBank mRNA | AM182458.1 | - | 7 - 8 | 1330 | 74 | |
| AY305003.1 | - | 7 - 8 | 1277 | 74 | ||
Set Membership: |
|