Search
Search

GALK| Gene Symbol: GALK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111534.4 | NP_187310.1 | 10 - 11 | 1441 | 86 | |
| GenBank mRNA | AY136356.1 | - | 10 - 11 | 1320 | 86 | |
| BT010374.1 | - | 10 - 11 | 1161 | 86 | ||
Set Membership: |
|