Search
Search

MFP2| Gene Symbol: MFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111566.4 | NP_187342.1 | 17 - 18 | 2336 | 104 | |
| GenBank mRNA | AF123254.1 | - | 17 - 18 | 2145 | 104 | |
| AY062621.1 | - | 17 - 18 | 2202 | 104 | ||
Set Membership: |
|