Search
Search

PIP5K6| Gene Symbol: PIP5K6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337754.1 | NP_001325991.1 | 7 - 8 | 2299 | 64 | |
| NM_111675.6 | NP_187453.1 | 7 - 8 | 2299 | 64 | ||
| GenBank mRNA | AK229597.1 | - | 7 - 8 | 2189 | 64 | |
| BT008595.1 | - | 7 - 8 | 2189 | 64 | ||
Set Membership: |
|