Search
Search

CSLG2| Gene Symbol: CSLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341637.1 | NP_001328486.1 | 2 - 3 | 373 | 123 | |
| NM_118532.4 | NP_567692.2 | 3 - 4 | 1073 | 123 | ||
| GenBank mRNA | AY070072.1 | - | 3 - 4 | 926 | 123 | |
| AY096446.1 | - | 3 - 4 | 838 | 123 | ||
| KJ138739.1 | - | 3 - 4 | 838 | 123 | ||
Set Membership: |
|