Search
Search

PWD| Gene Symbol: PWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341673.1 | NP_001328878.1 | 20 - 21 | 2505 | 106 | |
| NM_001341674.1 | NP_001328879.1 | 20 - 21 | 2509 | 106 | ||
| NM_118578.5 | NP_194176.3 | 19 - 20 | 2307 | 106 | ||
Set Membership: |
|