Search
Search

CLT2| Gene Symbol: CLT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203891.1 | NP_001190820.1 | 9 - 10 | 1167 | 82 | |
| NM_001341675.1 | NP_001329790.1 | 8 - 9 | 1382 | 82 | ||
| NM_001341676.1 | NP_001329789.1 | 8 - 9 | 1236 | 82 | ||
| NM_118579.4 | NP_194177.2 | 9 - 10 | 1177 | 82 | ||
| GenBank mRNA | BT029757.1 | - | 9 - 10 | 1133 | 82 | |
Set Membership: |
|