Search
Search

ZIM| Gene Symbol: ZIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203892.1 | NP_001190821.1 | 2 - 3 | 615 | 65 | |
| NM_118580.4 | NP_194178.1 | 2 - 3 | 635 | 65 | ||
| NM_179104.3 | NP_849435.1 | 3 - 4 | 517 | 65 | ||
| GenBank mRNA | AY086038.1 | - | 2 - 3 | 533 | 65 | |
Set Membership: |
|