Search
Search

QC| Gene Symbol: QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036639.3 | NP_001031716.1 | 4 - 5 | 553 | 113 | |
| NM_001036640.1 | NP_001031717.1 | 4 - 5 | 537 | 113 | ||
| NM_001341762.1 | NP_001329256.1 | 4 - 5 | 517 | 113 | ||
| NM_001341763.1 | NP_001329255.1 | 4 - 5 | 551 | 113 | ||
| NM_118704.2 | NP_567727.1 | 4 - 5 | 553 | 113 | ||
| GenBank mRNA | AK316976.1 | - | 4 - 5 | 486 | 113 | |
| AK318678.1 | - | 4 - 5 | 539 | 113 | ||
| BT001929.1 | - | 4 - 5 | 429 | 113 | ||
Set Membership: |
|