Search
Search

VATG3| Gene Symbol: VATG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341769.1 | NP_001328540.1 | 1 - 2 | 304 | 67 | |
| NM_118728.4 | NP_194325.1 | 1 - 2 | 283 | 67 | ||
| GenBank mRNA | AY063859.1 | - | 1 - 2 | 97 | 67 | |
| AY091217.1 | - | 1 - 2 | 87 | 67 | ||
Set Membership: |
|