Search
Search

ATK2| Gene Symbol: ATK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341838.1 | NP_001328096.1 | 15 - 16 | 2204 | 84 | |
| NM_118852.3 | NP_567768.1 | 16 - 17 | 2387 | 84 | ||
| GenBank mRNA | D21137.1 | - | 16 - 17 | 2231 | 84 | |
Set Membership: |
|