Search
Search

AT1G62305| Gene Symbol: AT1G62305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334027.1 | NP_001323374.1 | 3 - 4 | 676 | 135 | |
| NM_148618.5 | NP_683459.2 | 3 - 4 | 833 | 135 | ||
| GenBank mRNA | AY648315.1 | - | 3 - 4 | 562 | 135 | |
| AY924713.1 | - | 3 - 4 | 491 | 135 | ||
| BT020621.1 | - | 3 - 4 | 610 | 135 | ||
| BT021929.1 | - | 3 - 4 | 491 | 135 | ||
| KJ138956.1 | - | 3 - 4 | 491 | 135 | ||
Set Membership: |
|