Search
Search

CAMS1| Gene Symbol: CAMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334857.1 | NP_001321745.1 | 10 - 11 | 1744 | 90 | |
| NM_001334858.1 | NP_001321744.1 | 10 - 11 | 1744 | 90 | ||
| NM_001334859.1 | NP_001321746.1 | 10 - 11 | 1744 | 90 | ||
| NM_001334860.1 | NP_001321747.1 | 10 - 11 | 1737 | 90 | ||
| NM_148667.2 | NP_683508.1 | 11 - 12 | 1577 | 90 | ||
Set Membership: |
|