Search
Search

GATL6| Gene Symbol: GATL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036496.1 | NP_001031573.1 | 1 | 1288 | 64 | |
| NM_116445.2 | NP_192122.1 | 1 | 1452 | 64 | ||
| NM_178954.2 | NP_849285.1 | 1 | 1278 | 64 | ||
| GenBank mRNA | AK119044.1 | - | 1 | 1278 | 64 | |
| AK229344.1 | - | 1 | 1241 | 64 | ||
| BT000950.1 | - | 1 | 1007 | 64 | ||
Set Membership: |
|