Search
Search

ANT1| Gene Symbol: ANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337960.1 | NP_001327533.1 | 2 - 3 | 960 | 108 | |
| NM_112023.2 | NP_187796.1 | 2 - 3 | 960 | 108 | ||
| GenBank mRNA | AK227614.1 | - | 2 - 3 | 960 | 108 | |
| BT008302.1 | - | 2 - 3 | 889 | 108 | ||
Set Membership: |
|